MilliporeSigma
All Photos(1)

COM514239789

Combi-Blocks, Inc.

2,5-Bis(trifluoromethyl)benzoyl chloride

CAS Number:
Molecular Weight:
276.57
MDL number:

Iupac Name

2,5-bis(trifluoromethyl)benzoyl chloride

Inchi Code

1S/C9H3ClF6O/c10-7(17)5-3-4(8(11,12)13)1-2-6(5)9(14,15)16/h1-3H

InChI key

LRJNPOCQOZWIGR-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

95%

Country of Origin

CN

Product Link

http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4559

MSDS

http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4559

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service