Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

COMH6E40A3E9

Combi-Blocks, Inc.

Sultamicillin tosylate

Synonym(s):

({[(2S,5R)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl]carbonyl}oxy)methyl (2S,5R,6R)-6-{[(2R)-2-amino-2-phenylethanoyl]amino}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxy

Sign In to View Organizational & Contract Pricing.

Select a Size

1 G

$91.00

5 G

$279.50

10 G

$468.00

25 G

$936.00

100 G

$2,288.00

Vendor SKU: QA-8714-1G

$91.00


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
766.87
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C25H30N4O9S2.C7H8O3S/c1-24(2)17(28-13(30)10-14(28)39-24)22(33)37-11-38-23(34)18-25(3,4)40(35,36)21-16(20(32)29(18)21)27-19(31)15(26)12-8-6-5-7-9-12;1-6-2-4-7(5-3-6)11(8,9)10/h5-9,14-18,21H,10-11,26H2,1-4H3,(H,27,31);2-5H,1H3,(H,8,9,10)/t14-,15-,16-,17+,18+,21-;/m1./s1

Inchi Key

ZLGDCDFFYQXCHO-CGAOXQFVSA-N

Storage temp.

Refrigerated

Purity

98%

Country of Origin

CN

Looking for similar products? Visit Product Comparison Guide

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QA-8714


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service