Skip to Content
MilliporeSigma

COMH93D65AE0

Combi-Blocks, Inc.

5-Norbornene-2-ol

Sign In to View Organizational & Contract Pricing

Select a Size

Product COMH93D65AE0 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
110.16
MDL number:

Skip To

Iupac Name

(1R,4R)-bicyclo[2.2.1]hept-5-en-2-ol

Inchi Code

1S/C7H10O/c8-7-4-5-1-2-6(7)3-5/h1-2,5-8H,3-4H2/t5-,6+,7?/m0/s1

InChI key

MKOSBHNWXFSHSW-GFCOJPQKSA-N

storage temp.

Refrigerated

Purity

97%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?OR-3948

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?OR-3948


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library