Skip to Content
MilliporeSigma

COMH93D65C78

Combi-Blocks, Inc.

Dl-threo-3-phenylserine hydrate

Synonym(s):

(2S,3R)-2-amino-3-hydroxy-3-phenylpropanoic acid hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

1 G

$32.50

5 G

$104.00

25 G

$312.00

Vendor SKU: QA-2678-1G

$32.50


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
199.21
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C9H11NO3.H2O/c10-7(9(12)13)8(11)6-4-2-1-3-5-6;/h1-5,7-8,11H,10H2,(H,12,13);1H2/t7-,8+;/m0./s1

Inchi Key

DUGXBGWTWDCGNP-KZYPOYLOSA-N

Purity

98%

Country of Origin

CN

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QA-2678


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service