All Photos(1)

COMH93D67468

Combi-Blocks, Inc.

3-Morpholino-1,2-propanediol

CAS Number:
Molecular Weight:
161.2
MDL number:

Iupac Name

3-(4-morpholinyl)-1,2-propanediol

Inchi Code

1S/C7H15NO3/c9-6-7(10)5-8-1-3-11-4-2-8/h7,9-10H,1-6H2

InChI key

VZBNUEHCOOXOHR-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6108

MSDS

http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6108

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service