Skip to Content
MilliporeSigma

COMH9A887E22

Combi-Blocks, Inc.

2-Hydroxy-3,5-dinitrobenzoic acid hydrate

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product COMH9A887E22 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
246.13
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

2-hydroxy-3,5-dinitrobenzoic acid hydrate

Inchi Code

1S/C7H4N2O7.H2O/c10-6-4(7(11)12)1-3(8(13)14)2-5(6)9(15)16;/h1-2,10H,(H,11,12);1H2

InChI key

QRWHDJZDTLINBA-UHFFFAOYSA-N

Purity

98%

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?JV-7044

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?JV-7044


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library