All Photos(1)

ENA061957509

Enamine

3-[(3,4-difluorophenyl)sulfanyl]propanoic acid

CAS Number:
Molecular Weight:
218.22
MDL number:

mp

90-91

Iupac Name

3-[(3,4-difluorophenyl)sulfanyl]propanoic acid

Inchi Code

1S/C9H8F2O2S/c10-7-2-1-6(5-8(7)11)14-4-3-9(12)13/h1-2,5H,3-4H2,(H,12,13)

InChI key

OVLZSACZPZEUGH-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-14161

Melting Point

90-91

MSDS

https://www.enaminestore.com/catalog/EN300-14161

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service