Skip to Content
MilliporeSigma

ENA061957604

Enamine

(3-aminopropyl)(benzyl)methylamine

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
178.28
MDL number:
Marketplace Support
Need help or have questions about this product?
Contact Seller
Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

N~1~-benzyl-N~1~-methyl-1,3-propanediamine

Inchi Code

1S/C11H18N2/c1-13(9-5-8-12)10-11-6-3-2-4-7-11/h2-4,6-7H,5,8-10,12H2,1H3

InChI key

LZQYISZOMMPPIP-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-14111

pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-14111


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library