Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

ENA321763765

Enamine

6-methoxynaphthalene-2-carbothioamide

Synonym(s):

6-methoxy-2-naphthalenecarbothioamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ENA321763765 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C12H11O1N1S1
CAS Number:
Molecular Weight:
217.29
MDL number:

Skip To

Inchi Code

1S/C12H11NOS/c1-14-11-5-4-8-6-10(12(13)15)3-2-9(8)7-11/h2-7H,1H3,(H2,13,15)

Inchi Key

SZNBNJFTZHUGPB-UHFFFAOYSA-N

Purity

95

Storage temp.

RT

Country of Origin

UA

Physical Form

powder


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library