Skip to Content
MilliporeSigma

ENA341292926

Enamine

2,2,2-trifluoroethyl N-(3-chloro-4-fluorophenyl)carbamate

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
271.6
MDL number:
Marketplace Support
Need help or have questions about this product?
Contact Seller
Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

2,2,2-trifluoroethyl 3-chloro-4-fluorophenylcarbamate

Inchi Code

1S/C9H6ClF4NO2/c10-6-3-5(1-2-7(6)11)15-8(16)17-4-9(12,13)14/h1-3H,4H2,(H,15,16)

InChI key

KPBAQGXPKDBHJV-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-88276

MSDS

https://www.enaminestore.com/catalog/EN300-88276


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library