Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA369383020

Enamine

bis(N-propylguanidine); sulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
300.38
MDL number:
Product ENA369383020 is currently not available for purchase. Contact Technical Service

Iupac Name

sulfuric acid compound with N-propylguanidine (1:2)

Inchi Code

1S/2C4H11N3.H2O4S/c2*1-2-3-7-4(5)6;1-5(2,3)4/h2*2-3H2,1H3,(H4,5,6,7);(H2,1,2,3,4)

InChI key

VZKCJJUATTWLTC-UHFFFAOYSA-N

storage temp.

RT

Salt Data

O=S(=O)(O)O

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-104017

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-104017


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library