MilliporeSigma
All Photos(1)

ENA370563227

Enamine

2-methylbutane-1-thiol

CAS Number:
Molecular Weight:
104.22
MDL number:

Iupac Name

2-methyl-1-butanethiol

Inchi Code

1S/C5H12S/c1-3-5(2)4-6/h5-6H,3-4H2,1-2H3

InChI key

WGQKBCSACFQGQY-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-105388

MSDS

https://www.enaminestore.com/catalog/EN300-105388

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service