All Photos(1)

ENA371463751

Enamine

N-methyl-N-propylsulfamoyl chloride

Molecular Weight:
171.65

Iupac Name

methyl(propyl)sulfamoyl chloride

Inchi Code

1S/C4H10ClNO2S/c1-3-4-6(2)9(5,7)8/h3-4H2,1-2H3

InChI key

MHIVSJQHEDZPPG-UHFFFAOYSA-N

storage temp.

RT

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service