Skip to Content
MilliporeSigma

ENA371464270

Enamine

1-benzyl-4-chlorophthalazine

Synonym(s):

1-benzyl-4-chlorophthalazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H11N2Cl1
CAS Number:
Molecular Weight:
254.72
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H11ClN2/c16-15-13-9-5-4-8-12(13)14(17-18-15)10-11-6-2-1-3-7-11/h1-9H,10H2

Inchi Key

COIJTZLLJUXFNV-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Melting Point

147-152





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library