MilliporeSigma
All Photos(1)

ENA410543529

Enamine

2-(diethylamino)acetonitrile

CAS Number:
Molecular Weight:
112.17
MDL number:

Iupac Name

(diethylamino)acetonitrile

Inchi Code

1S/C6H12N2/c1-3-8(4-2)6-5-7/h3-4,6H2,1-2H3

InChI key

LVPZSMIBSMMLPI-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-17594

Flash Point

53

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

H226,H301,H311,H316,H319

MSDS

https://www.enaminestore.com/catalog/EN300-17594

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service