All Photos(1)

ENA429359898

Enamine

[5-(4-fluorophenyl)-4-methyl-4H-1,2,4-triazol-3-yl]methanol

CAS Number:
Molecular Weight:
207.21
MDL number:

Iupac Name

[5-(4-fluorophenyl)-4-methyl-4H-1,2,4-triazol-3-yl]methanol

Inchi Code

1S/C10H10FN3O/c1-14-9(6-15)12-13-10(14)7-2-4-8(11)5-3-7/h2-5,15H,6H2,1H3

InChI key

CRJCOEFQLBEIHM-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-188937

MSDS

https://www.enaminestore.com/catalog/EN300-188937

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service