MilliporeSigma
All Photos(1)

ENA947386502

Enamine

3-(dimethylamino)propanenitrile

CAS Number:
Molecular Weight:
98.15
MDL number:

Iupac Name

3-(dimethylamino)propanenitrile

Inchi Code

1S/C5H10N2/c1-7(2)5-3-4-6/h3,5H2,1-2H3

InChI key

MTPJEFOSTIKRSS-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-18465

Flash Point

63

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-18465

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service