Skip to Content
MilliporeSigma

ENAH304581E9

Enamine

1,1-dimethyl 2-methylcyclobutane-1,1-dicarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

Product ENAH304581E9 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
186.21
MDL number:

Skip To

Iupac Name

dimethyl 2-methyl-1,1-cyclobutanedicarboxylate

Inchi Code

1S/C9H14O4/c1-6-4-5-9(6,7(10)12-2)8(11)13-3/h6H,4-5H2,1-3H3

InChI key

OQTZRIUJDWUIFM-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-128767

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library