Sign In to View Organizational & Contract Pricing
Select a Size
1 G
$1,355.90
2.5 G
$2,655.90
5 G
$3,929.90
10 G
$5,827.90
Vendor SKU: EN300-219115-1G
$1,355.90
Ships in 30 days - from Aldrich Partner.
About This Item
Skip To
Iupac Name
sulfuric acid compound with 2,4,5-pyrimidinetriamine (1:1)
Inchi Code
1S/C4H7N5.H2O4S/c5-2-1-8-4(7)9-3(2)6;1-5(2,3)4/h1H,5H2,(H4,6,7,8,9);(H2,1,2,3,4)
InChI key
VQKFIHPLERCHFG-UHFFFAOYSA-N
Salt Data
OS(=O)(=O)O
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-219115
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service