Skip to Content
MilliporeSigma

ENAH3049BB01

Enamine

3-[(1-benzyl-1H-pyrazol-4-yl)oxy]-4-hydroxy-1lambda6-thiolane-1,1-dione

Synonym(s):

3-((1-benzyl-1H-pyrazol-4-yl)oxy)-4-hydroxytetrahydrothiophene 1,1-dioxide

Sign In to View Organizational & Contract Pricing.

Select a Size


Inchi Code

1S/C14H16N2O4S/c17-13-9-21(18,19)10-14(13)20-12-6-15-16(8-12)7-11-4-2-1-3-5-11/h1-6,8,13-14,17H,7,9-10H2

Inchi Key

MQWWTKOTVRSWRF-UHFFFAOYSA-N

Country of Origin

UA


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library