Sign In to View Organizational & Contract Pricing
Select a Size
Product ENAH304E4687 is currently not available for purchase. Contact Technical Service
Skip To
Iupac Name
2-amino-4-methyl-5-(methylsulfonyl)pentanamide
Inchi Code
1S/C7H16N2O3S/c1-5(4-13(2,11)12)3-6(8)7(9)10/h5-6H,3-4,8H2,1-2H3,(H2,9,10)
InChI key
WQRVTKZSBVBUAM-UHFFFAOYSA-N
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-1300039
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.