All Photos(1)

ENAH90822220

Enamine

2-ethyl-3-methylbutanal

Molecular Weight:
114.19
MDL number:

Iupac Name

2-ethyl-3-methylbutanal

Inchi Code

1S/C7H14O/c1-4-7(5-8)6(2)3/h5-7H,4H2,1-3H3

InChI key

SHGPBDQRELYPLO-UHFFFAOYSA-N

storage temp.

+4

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-2971164

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

H317,H411

MSDS

https://www.enaminestore.com/catalog/EN300-2971164

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service