Iupac Name
7-(aminomethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Inchi Code
1S/C9H9N3O2/c10-4-6-1-2-12-5-7(9(13)14)11-8(12)3-6/h1-3,5H,4,10H2,(H,13,14)
InChI key
OOKZSXRUVNOGEV-UHFFFAOYSA-N
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-27722203
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service