Skip to Content
MilliporeSigma

ENAH9926CE13

Enamine

[(2-amino-1,3-benzothiazol-6-yl)sulfanyl]formonitrile

Synonym(s):

2-amino-1,3-benzothiazol-6-yl thiocyanate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H5N3S2
CAS Number:
Molecular Weight:
207.28
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H5N3S2/c9-4-12-5-1-2-6-7(3-5)13-8(10)11-6/h1-3H,(H2,10,11)

Inchi Key

FVNSRFMQXKMHTQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95

Storage temp.

RT

Country of Origin

UA

Physical Form

powder

Melting Point

195-197



hstatements

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library