Iupac Name
λ²-ruthenium(2+) tris(2,2'-bipyrimidine) dichloride
Inchi Code
1S/3C8H6N4.2ClH.Ru/c3*1-3-9-7(10-4-1)8-11-5-2-6-12-8;;;/h3*1-6H;2*1H;/q;;;;;+2/p-2
InChI key
DVHNMJRXSLLELA-UHFFFAOYSA-L
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-53156406
MSDS
https://www.enaminestore.com/catalog/EN300-53156406
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.