FLUH99CD232A
Fluorochem (Preferred Partner)
NALPHA-(2,4-DINITROPHENYL)-L-ARGININE
Iupac Name
(2S)-5-{[amino(imino)methyl]amino}-2-(2,4-dinitroanilino)pentanoic acid
Inchi Code
1S/C12H16N6O6/c13-12(14)15-5-1-2-9(11(19)20)16-8-4-3-7(17(21)22)6-10(8)18(23)24/h3-4,6,9,16H,1-2,5H2,(H,19,20)(H4,13,14,15)/t9-/m0/s1
InChI key
GZJXZYUXRVBOAH-VIFPVBQESA-N
storage temp.
Ambient Storage
Purity
98%
Product Link
https://fluorochem.co.uk/product/F800906
Signal Word
Warning
Hazard Statements
MSDS
https://sds.fluorochem.co.uk/eng/sds_F800906_en.pdf
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
