MilliporeSigma
All Photos(1)

1509017

Foxx Life Sciences

Foxx Life Sciences Borosil® Reagent Bottles Amber

Amber Reagent Bottles Plain Narrow Mouth, Graduated I/C Glass Stopper 125 mL , 50/CS

Synonym(s):
Bottles

capacity

125 mL

feature

with cap

material

Bottle: borosilicate glass 3.3
Cap: plastic

manufacturer/tradename

Foxx Life Sciences

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
150901315090161509030
capacity

125 mL

capacity

60 mL

capacity

100 mL

capacity

2000 mL

feature

with cap

feature

with cap

feature

with cap

feature

with cap

material

Bottle: borosilicate glass 3.3

material

Bottle: borosilicate glass 3.3, Cap: plastic

material

Bottle: borosilicate glass 3.3, Cap: plastic

material

Bottle: borosilicate glass 3.3, Cap: plastic

General description

Borosil® Amber Reagent Bottles, graduated with interchangeable amber flat head stoppers are chemically resistant and mechanically strong. These narrow mouth storage vessels have a sturdy ground neck and come standard with graduations and an enameled area for easy marking. The amber color is highly durable and protects light-sensitive media from light radiation. All components are autoclavable and sterilizable. High resistance to thermal shock up to 140°C. Complies with IS 1388 (Part II), EN ISO 4796 - 2. *60mL & 125mL not covered in IS and EN ISO standards.


SPECIFICATIONS:

  • Material: Glass
  • Bottle: 125 mL
  • Overall Dimension: 56mm X 120mm
  • Neck Type: Grounded Neck I/C Stopper
  • Neck Size: 19/26
  • Borosilicate Glass 3.3
  • Graduation Range: Type I
  • Quantity: Case of 50

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA