Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1509029

Foxx Life Sciences

Foxx Life Sciences Borosil® Reagent Bottles Amber

Bottles, Reagent, Amber, with Stoppers, 1,000mL, 29/32, 5/CS

Synonym(s):

Bottles

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$836.74

$836.74


Available to ship onFebruary 04, 2025Details



Select a Size

Change View
5 EA
$836.74

About This Item

PDF Icon
Shipped & Sold By Foxx Life Sciences
A Trusted Marketplace Seller
PDF Icon
Have a question?

$836.74


Available to ship onFebruary 04, 2025Details


capacity

1000 mL

feature

with cap

material

Bottle: borosilicate glass 3.3
Cap: plastic

manufacturer/tradename

Foxx Life Sciences

General description

Borosil® Amber Reagent Bottles, graduated with interchangeable amber flat head stoppers are chemically resistant and mechanically strong. These narrow mouth storage vessels have a sturdy ground neck and come standard with graduations and an enameled area for easy marking. The amber color is highly durable and protects light-sensitive media from light radiation. All components are autoclavable and sterilizable. High resistance to thermal shock up to 140°C. Complies with IS 1388 (Part II), EN ISO 4796 - 2. *60mL & 125mL not covered in IS and EN ISO standards.


SPECIFICATIONS:

  • Material: Glass
  • Bottle: 1000 mL
  • Overall Dimension: 101mm X 230mm
  • Neck Type: Grounded Neck I/C Stopper
  • Neck Size: 29/32
  • Borosilicate Glass 3.3
  • Graduation Range: Type I
  • Quantity: Case of 5

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library