Skip to Content
MilliporeSigma

GDFH9BC739CC

Goodfellow

Gold Foil (Light Tight)

Gold Foil (Light Tight),Formula: AuPercentage Purity: 99.99%Temper: As RolledThickness: 0.25mmLength 1: 25mmLength 2: 50mmCAS Number: 7440-57-5UOM Code: 613-221-86Legacy Code: AU000370Distributor Code: GF61322186

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
0
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Shipping Temperature

Ambient

Purity

99.99%

Storage temp.

Ambient

Country of Origin

US

Physical Form

Solid

Melting Point

1064.4 C

Density

19.32g/cm³

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service