All Photos(1)

MAN182107160

Manchester Organics

3,5-Diiodo-4-pyridone-1-acetic acid

CAS Number:
Molecular Weight:
404.93
MDL number:

Iupac Name

(3,5-diiodo-4-oxo-1(4H)-pyridinyl)acetic acid

Inchi Code

1S/C7H5I2NO3/c8-4-1-10(3-6(11)12)2-5(9)7(4)13/h1-2H,3H2,(H,11,12)

InChI key

PVBALTLWZVEAIO-UHFFFAOYSA-N

Purity

0.97

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/F16348

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service