Skip to Content
MilliporeSigma

MAT390509603

Matrix Scientific

Methyl 2-chloropropanoate

Synonym(s):

methyl 2-chloropropanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

10 G

$16.80

25 G

$22.80

Vendor SKU: 098291-10G

$16.80


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
122.55
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

density

1.075

bp

132-133

mp

-10

Inchi Code

1S/C4H7ClO2/c1-3(5)4(6)7-2/h3H,1-2H3

Inchi Key

JLEJCNOTNLZCHQ-UHFFFAOYSA-N

Purity

95%

Country of Origin

US

Flash Point

38

MSDS

https://www.matrixscientific.com/098291.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service