MilliporeSigma
All Photos(1)

Documents

02-0111-03

Millipore

Erenna 10X System Buffer w/ Proclin

Sign Into View Organizational & Contract Pricing

eCl@ss:
32169090

form

liquid

Quality Level

manufacturer/tradename

Singulex®

technique(s)

single molecule counting: suitable

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
B642934062BC40-50021
vibrant-m

B6429

Casein Blocking Buffer 10x

technique(s)

single molecule counting: suitable

technique(s)

Southern blotting: suitable

technique(s)

ELISA: suitable

technique(s)

multiplexing: suitable

Quality Level

300

Quality Level

200

Quality Level

-

Quality Level

-

manufacturer/tradename

Singulex®

manufacturer/tradename

-

manufacturer/tradename

BioControl Systems TRANSIA® SA0180

manufacturer/tradename

Milliplex®

General description

The Erenna Instruments require 10x Wash Buffer to serve as the delivery medium to transport the sample to the instrument′s optics.

Application

Erenna 10X System Buffer w/ Proclin has been used as a wash buffer for Erenna Singulex SMCTM immunoassay.
For use with Erenna System

Storage and Stability

Store at room temperature.

Legal Information

SMC is a registered trademark of Singulex

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B - Skin Sens. 1

Storage Class

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Brain Biomarkers: Follow-Up of RNA Expression Discovery Approach: CSF Assays for Neurogranin, SNAP-25, and VILIP-1
FABAD Journal of Pharmaceutical Sciences, 181-221 (2021)
Correction: Shedding a new light on Huntington's disease: how blood can both propagate and ameliorate disease pathology.
Marie Rieux et al.
Molecular psychiatry, 26(9), 5464-5464 (2020-07-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service