Skip to Content
MilliporeSigma
All Photos(1)

Documents

02-0409-00

Millipore

Tris Buffered Saline

600mM NaCI/MoGoMAK/EDTA/0.1%Tx-100

Sign Into View Organizational & Contract Pricing

UNSPSC Code:
12352200
NACRES:
NA.25

product name

TBS/600mM NaCI/MoGoMAK/EDTA/0.1%Tx-100,

Quality Level

hcodes

Hazard Classifications

Aquatic Chronic 3

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service