Documents

03-0122-00

Millipore

SMC optimization kit, Bead-Based Assay Buffer Optimization Kit

eCl@ss:
32161000
Pricing and availability is not currently available.

Quality Level

mfr. no.

SMC

application(s)

ELISA: suitable
single molecule counting: suitable

General description

Take advantage of Single Molecule Counting (SMC) technology while developing your own application-specific immunoassays in bead format. This kit provides everything you need to build your own high performing SMC immunoassay. It includes buffers and protocol to optimize conditions for Bead Based assay development. The reagents are suitable for up to 65 optimization experiments.

Application

Up to 65 optimization experiments.
Optimize your bead-based assay buffer and move beyond the traditional ELISA with TMC="SMC" Technology.

Components

Optimization Assay Buffer A
Optimization Assay Buffer B
Optimization Assay Buffer C

Legal Information

SMC is a trademark of Singulex

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.