Skip to Content
MilliporeSigma

03-0219-00

SMC® Human IL-22 Binding Protein High Sensitivity Immunoassay Kit

new

SMC® advanced ultrasensitive immunoassay technology

Synonym(s):

IL-22 inhibitory protein immunoassay, IL-22 receptor antagnoist assay, high sensitivity IL-22BP assay, interleukin-22 binding protein high sensitivity assay, ultra-sensitive IL-22BP

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

usage

 kit sufficient for 96 wells

quality

200

species reactivity

human

manufacturer/tradename

SMC®

assay range

linearity: 113%
(Serum)

linearity: 115%
(Plasma)

recovery range: 116%
(Serum)

recovery range: 98%
(Plasma)

sensitivity: 1.95-7.81 pg/mL
(Refer to CoA for lot-specific LLOQ)

standard curve range: 0.98-1,000 pg/mL

technique(s)

single molecule counting: suitable

input

plasma (K2EDTA)
serum

shipped in

wet ice

storage temp.

2-8°C

General description

IL-22 binding protein (IL-22BP) is a soluble protein that regulates the immune response by binding to IL-22 and preventing it from interacting with its receptor on target cells. This action serves as a negative regulator, helping maintain a balanced immune response and preventing excessive inflammation. IL-22BP may have therapeutic potential in treating autoimmune and inflammatory diseases by modulating IL-22 signaling.

Application

For ultrasensitive detection of IL-22BP in human serum and plasma

Features and Benefits

Features
  • Robustness and Reproducibility: Lot-matched components ensure consistent assay performance across experiments, reducing variability and enhancing data reliability.
  • Enhanced Detection Limits: Capable of detecting ultra-low analyte levels in the picogram and femtogram range, enabling the study of low-abundance biomarkers critical in early-stage research.
  • Simplified Workflow Compatibility: Designed to align with standard assay workflows, including ELISA, minimizing the need for additional training or protocol adjustments.
  • Certified Data Assurance: Each kit includes a Certificate of Analysis detailing validated assay characteristics, ensuring traceability and quality control for regulatory compliance.
  • Wide Dynamic Range: Accurately quantifies analyte concentrations across a broad range, supporting both high and low abundance detection in complex samples.
  • Automation Ready: Compatible with high-throughput systems, enabling seamless integration into automated workflows, reducing manual handling errors, and increasing efficiency.
Benefits
  • Understanding Mechanisms: By accurately measuring IL-22BP levels, you can explore its role in immune responses and pathophysiology, enhancing your understanding of disease mechanisms.
  • Monitoring Disease Models: This ultrasensitive assay facilitates tracking of IL-22BP levels over time in your samples, helping you gain insights for research into disease progression and treatment effects.
  • Early Detection: With enhanced sensitivity, this assay allows you to detect low concentrations of IL-22BP, enabling early identification of inflammatory and autoimmune conditions in your studies.
  • Integrated Analysis: Having both IL-22BP and IL-22 assays on the same platform allows for streamlined data collection and analysis, enabling you to correlate IL-22 levels with its binding protein in a single workflow.

Legal Information

SMC is a registered trademark of Singulex

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service