description
Kit sufficient for 96 wells
Quality Level
quality
200
species reactivity
human
manufacturer/tradename
SMC®
assay range
linearity: 113%
(plasma)
linearity: 115%
(serum)
recovery range: 116%
(plasma)
recovery range: 127%
(serum)
sensitivity: 0.1-0.91 pg/mL
(Refer to CoA for lot-specific LLOQ)
standard curve range: 0.03-2,000 pg/mL
technique(s)
single molecule counting: suitable
input
sample type serum
sample type plasma (K2EDTA)
shipped in
wet ice
storage temp.
2-8°C
General description
Application
Features and Benefits
- Robustness and Reproducibility: Lot-matched components ensure consistent assay performance across experiments, reducing variability and enhancing data reliability.
- Enhanced Detection Limits: Capable of detecting ultra-low analyte levels in the picogram and femtogram range, enabling the study of low-abundance biomarkers critical in early-stage research.
- Simplified Workflow Compatibility: Designed to align with standard assay workflows, including ELISA, minimizing the need for additional training or protocol adjustments.
- Certified Data Assurance: Each kit includes a Certificate of Analysis detailing validated assay characteristics, ensuring traceability and quality control for regulatory compliance.
- Wide Dynamic Range: Accurately quantifies analyte concentrations across a broad range, supporting both high and low abundance detection in complex samples.
- Automation Ready: Compatible with high-throughput systems, enabling seamless integration into automated workflows, reducing manual handling errors, and increasing efficiency.
- Understanding Immune Regulation: Ultrasensitive measurement of CTLA-4 allows researchers to explore immune modulation mechanisms, contributing to a deeper understanding of immune responses in various biological contexts.
- Modeling Disease Mechanisms: By measuring CTLA-4 levels, researchers can investigate its role in disease models, providing insights into the pathophysiology of conditions such as cancer and autoimmune diseases.
- Evaluating Therapeutic Strategies: In pre-clinical studies, monitoring CTLA-4 levels can help assess the effectiveness of novel immunotherapies or other treatments aimed at modulating immune responses.
- Biomarker Discovery: The ability to measure CTLA-4 sensitively may aid in identifying potential biomarkers for immune-related conditions, enhancing predictive models in research.
- Facilitating Drug Development: Understanding the dynamics of CTLA-4 in pre-clinical models can inform drug development processes, guiding the design of more effective therapies targeting immune checkpoints.
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B - STOT RE 2 Inhalation
target_organs
Respiratory Tract
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service