1.00633

Millipore

Acrylamide ready-to-use solution 40%

for electrophoresis

storage conditions

Store at +2°C to +8°C.

Quality Level

density

1.031 g/cm3 at 20 °C

storage temp.

2-8°C

Analysis Note

Assay (by acidimetry following saponification): 39.0 - 41.0 %
Appearance (colour): almost colourless
Appearance (clearness): almost clear
Free acid (as acrylic acid): ≤ 0.001 %
Spec. Absorptivity A 1%/1cm (290 nm; 1 %; 1 cm; water): ≤ 0.1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.