MilliporeSigma
All Photos(2)

1.04005

SAFC

Paraformaldehyde

EMPROVE® ESSENTIAL, DAC

Synonym(s):
Paraformaldehyde
Linear Formula:
(CH2O)n
CAS Number:
MDL number:

Agency

DAC

Quality Level

vapor pressure

1.93 hPa ( 25 °C)

product line

EMPROVE® ESSENTIAL

assay

95-100.5%

form

solid

autoignition temp.

300 °C

expl. lim.

7-73 % (v/v)

technique(s)

immunofluorescence: suitable

pH

5.5 (20 °C in H2O, saturated solution)

mp

100-130 °C

density

1.4 g/cm3 at 20 °C

application(s)

pharmaceutical

storage temp.

2-25°C

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Sol. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials.

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service