Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.04481

Supelco

Mercury(II) sulfate

EMPLURA®

Synonym(s):

Mercury(II) sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HgO4S
CAS Number:
Molecular Weight:
296.65
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-992-5
Pricing and availability is not currently available.

product line

EMPLURA®

assay

≥99.0% (complexometric)

form

solid

autoignition temp.

>450 °C

pH

1 (20 °C, 50 g/L in H2O)

density

6.47 g/cm3 at 20 °C

bulk density

670 kg/m3

anion traces

chloride (Cl-): ≤0.003%

cation traces

Fe: ≤0.005%
Hg+: ≤0.1%

storage temp.

2-30°C

InChI

1S/Hg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

DOBUSJIVSSJEDA-UHFFFAOYSA-L

Analysis Note

Assay (complexometric): ≥ 99.0 %
Identity: passes test
Chloride (Cl): ≤ 0.003 %
Fe (Iron): ≤ 0.005 %
Mercury (I) (as Hg): ≤ 0.1 %
Substances not reducing by formic acid (residue on ignition): ≤ 0.1 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service