MilliporeSigma
All Photos(1)

Documents

1.06197

Millipore

2-Morpholinoethanesulfonic acid sodium salt (MES Na)

buffer substance

Synonym(s):
2-Morpholinoethanesulfonic acid sodium salt (MES Na)
CAS Number:
Molecular Weight:
217.22
MDL number:
EC Index Number:
275-203-2

form

powder or crystals

pH

8.9-10.1 (20 °C, 10 g/L in H2O)

storage temp.

2-30°C

InChI

1S/C6H13NO4S.Na/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);/q;+1/p-1

InChI key

IRHWMYKYLWNHTL-UHFFFAOYSA-M

General description

buffer substance

Application

2-Morpholinoethanesulfonic acid sodium salt (MES Na) buffer substance. CAS 71119-23-8, pH 8.9 - 10.1 (10 g/l, H₂O, 20 °C).

Analysis Note

Assay (alkalimetric, calculated on dried substance)≥ 99.0 %Identity (IR-spectrum)passes testAppearancewhite to almost white, fine powder or colorless crystalsAppearance of solution (0.5 mol/l, water)clear and colorless.UV-Absorption (260 nm; 0.5 mol/l; 1 cm; water)≤ 0.05UV-Absorption (280 nm; 0.5 mol/l; 1 cm; water)≤ 0.05Chloride (Cl)≤ 50 ppmSulphate (SO₄)≤ 50 ppmHeavy metals (as Pb)≤ 5 ppmCa (Calcium)≤ 50 ppmCd (Cadmium)≤ 5 ppmCo (Cobalt)≤ 5 ppmCr (Chromium)≤ 5 ppmCu (Copper)≤ 5 ppmFe (Iron)≤ 10 ppmK (Potassium)≤ 50 ppmMg (Magnesium)≤ 10 ppmMn (Manganese)≤ 5 ppmNi (Nickel)≤ 5 ppmPb (Lead)≤ 5 ppmZn (Zinc)≤ 5 ppmDNases (Exo- and endonucleases)non detectableRNasesnon detectableProteasesnon detectableNicking activity≤ 10 %Loss on Drying (105°C)≤ 1.0 %Biological buffer, pKa at 20 °C6.1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 5

1 of 5

MES monohydrate BioUltra, for molecular biology, ≥99.5% (T)

Sigma-Aldrich

69889

MES monohydrate

MES hydrate BioPerformance Certified, suitable for cell culture, ≥99.5%

Sigma-Aldrich

M2933

MES hydrate

Oxalic acid puriss. p.a., anhydrous, ≥99.0% (RT)

Sigma-Aldrich

75688

Oxalic acid

MES hydrate ≥99.5% (titration)

Sigma-Aldrich

M8250

MES hydrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service