Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09229

Millipore

Battery pack NIMH

for use with MAS-100®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403

packaging

pkg of 1 kit

compatibility

for use with MAS-100®

storage temp.

no temp limit

General description

Battery pack NIMH is an additional accessory for MAS-100® air sampler. The MAS-100® air sampler is designed to monitor air microbiology in cleanrooms and hygienically ultra-sensitive areas. Its high recovery rate is based on the Andersen impaction principle. An integrated airflow sensor compensates for differences in pressure and temperature in the environment. The Nickel-metal hydroxide batteries are rechargeable and environment friendly batteries with no memory effect. A complete recharging cycle takes about 9 hours. With the batteries fully charged, the instrument has a total capacity of approximately 18,000 liters of air, equivalent to about 180 readings @ 100 liters.

Application

Battery pack NIMH is an additional accessory for MAS-100® Air Sampler that are rechargeable and provide power supply. Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.

Legal Information

MAS-100 is a registered trademark of MBV AG

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service