MilliporeSigma
All Photos(1)

Documents

1.10236

Millipore

Salmonella enrichment broth acc. to RAPPAPORT

for the selective enrichment of Salmonella

Sign Into View Organizational & Contract Pricing

Quality Level

form

powder (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

5.8-6.2 (25 °C, 32.7 g/L in H2O, after autoclaving)

solubility

32.7 g/L

application(s)

clinical
microbiology

storage temp.

15-25°C

suitability

Salmonella spp.

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
1.107561.164341.07666
vibrant-m

1.10756

GN enrichment broth acc. to HAJNA

suitability

Salmonella spp.

suitability

Shigella spp., enterobacteriaceae

suitability

Yersinia spp.

suitability

Salmonella spp.

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

manufacturer/tradename

GranuCult®

manufacturer/tradename

GranuCult®

manufacturer/tradename

-

manufacturer/tradename

NutriSelect® Prime

application(s)

clinical
microbiology

application(s)

clinical
microbiology

application(s)

clinical
food and beverages
microbiology

application(s)

microbiology, pharmaceutical

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

General description

for the selective enrichment of Salmonella

Analysis Note

Appearance (clearness)turbidAppearance (colour)dark bluepH-value (25 °C)5.8 - 6.2Inoculum (Escherichia coli ATCC 25922 (WDCM 00013))about 99 %Inoculum (Salmonella typhimurium ATCC 14028 (WDCM 00031))about 1 %Enrichment from a mixed inoculum after 6 hours (Escherichia coli ATCC 25922 (WDCM 00013))Enrichment from a mixed inoculum after 6 hours (Salmonella typhimurium ATCC 14028 (WDCM 00031))Enrichment from a mixed inoculum after 24 hours (Escherichia coli ATCC 25922 (WDCM 00013))≤ 10 %Enrichment from a mixed inoculum after 24 hours (Salmonella typhimurium ATCC 14028 (WDCM 00031))≥ 90 %Enrichment of Salmonella from a mixed inoculum;
Incubation: 24 hrs.; 35 °C; detection with BROLACIN agar.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service