Recommended Products
form
crystalline powder (Fine)
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
brown-orange
SMILES string
[Zn+2].[Cl-].[Cl-].[Cl-].N(C)(C)c1cc2nc3c(cc2cc1)ccc(c3)N(C)C.[H+]
InChI
1S/C17H19N3.3ClH.Zn/c1-19(2)14-7-5-12-9-13-6-8-15(20(3)4)11-17(13)18-16(12)10-14;;;;/h5-11H,1-4H3;3*1H;/q;;;;+2/p-2
InChI key
VADJQOXWNSPOQA-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Specifications:
Molar absorptivity Em (490 nm, water): ≥ 36500
Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service