Sign In to View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
NACRES:
NA.21
Skip To
material
copper
packaging
pack of 1 unit
manufacturer/tradename
USB-C Cable Sirius
technique(s)
: suitable using airmonitoring
compatibility
MAS-100® Sirius®
Related Categories
General description
The USB-C Cable is included with the MAS-100 Sirius but can also be ordered separately.
This cable is identified as USB Type C 2.0 PD fast charging, supporting up to 60W (20V, 3A) and has a length of 1.5 meters.
This cable is identified as USB Type C 2.0 PD fast charging, supporting up to 60W (20V, 3A) and has a length of 1.5 meters.
Application
The USB-C cable is used for charging as well as for connecting to a PC to access the instrument-integrated software via a browser.
Legal Information
MAS-100 is a registered trademark of MBV AG
MAS-100 Sirius is a registered trademark of MBV AG