Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

125144

Millipore

RoboCol Fractogel® TMAE Medcap (M) 0.2 ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

ligand

(Trimethylammoniumethyl)

material

polypropylene housing

product line

Fractogel®

parameter

8 bar max. pressure
~180 cm/hr max. flow rate

volume

0.2 mL

matrix active group

Methacrylate

mean particle size

40-90 μm

capacity

~150  mg binding capacity (BSA/mg of resin)

application(s)

plasma purification
recombinant protein
vaccine development

separation technique

anion exchange

General description

Strong anion exchange Fractogel® TMAE Medcap (M) resin in a convenient pre-packed 0.2 ml RoboColumn used in 96-well plate automated liquid handling systems

Features and Benefits

RoboCol Fractogel® TMAE MedCap (M) 0.2 ml enables:
  • Excellent binding at high flow rates
  • Excellent pressure-flow behavior
  • Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution

Packaging

pkg of 8 pieces

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

95.0 °F

flash_point_c

35 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service