MilliporeSigma
All Photos(1)

1.30104

Millipore

HY-LiTE® Rinse Solution

HY-LiTE

Sign Into View Organizational & Contract Pricing

Select a Size

Synonym(s):
Water, Deionized water, Water
Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:

vapor pressure

23 hPa ( 20 °C)

Quality Level

form

liquid

packaging

pkg of 50 vials

refractive index

n20/D 1.34 (lit.)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

application(s)

cosmetics
food and beverages
industrial hygiene

storage temp.

2-8°C

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
1.153338.483331.01262
HY-LiTE® Rinse Solution HY-LiTE

Millipore

1.30104

HY-LiTE® Rinse Solution

Water for chromatography (LC-MS Grade) LiChrosolv®

Supelco

1.15333

Water

Di water for synthesis

Sigma-Aldrich

8.48333

Di water

Water Ultrapur

Supelco

1.01262

Water

form

liquid

form

liquid

form

(Colorless Liquid)

form

liquid

refractive index

n20/D 1.34 (lit.)

refractive index

n20/D 1.34 (lit.)

refractive index

n20/D 1.34 (lit.)

refractive index

n20/D 1.34 (lit.)

bp

100 °C (lit.)

bp

100 °C (lit.)

bp

100 °C (lit.)

bp

100 °C (lit.)

mp

0 °C (lit.)

mp

0 °C (lit.)

mp

0 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

General description

Rinse Solution is intended for use with HY-LiTE® 2/2A luminometers and ATP pens/test kits for hygiene monitoring of surfaces in food and beverages production. It is an easy-to-use, rapid and reliable test system to assess the quality of processing equipment, surfaces, and other environments. The swab is dipped in the rinse solution after swabbing the area of interest of a typical flat surface in order to transfer the sample. The white stick of the pen can then draw a sample of the rinse solution where it can then be read within the luminometer.

Application

HY-LiTE® Rinse solution is designed for use with HY-LiTE® sampling pens for hygiene monitoring of surfaces and other product contact areas in food and beverages production plants.

Linkage

SKU 1.30104.0002 replaced by 1.30104.0001

Legal Information

HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service