Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.44132

Millipore

Lithium-ion Battery

for use with APC Ergo Touch Pro 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104906
NACRES:
NA.74

application(s)

pharmaceutical

compatibility

for use with APC Ergo Touch Pro 2

storage temp.

room temp

General description

942561

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service