Skip to Content

14422AM-M

YM Discs 100K NMWL 44.5MM 10PK

Synonym(s):

YM Discs 100K NMWL 44.5MM 10PK

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
40161507
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ultracel® 100 kDa Ultrafiltration Discs, Chemistry: Regenerated Cellulose (RC) Filter Diameter: 44.5 mm Amicon Bioseparations

Quality Segment

Legal Information

ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library