1.46427
MacConkey Agar Contact
55 mm Contact Plates, for isolation and detection of Enterobacteriaceae -Room Temperature storage, accordance with EP
Select a Size
$83.00
Select a Size
About This Item
$83.00
Recommended Products
material
polystyrene plate
Quality Level
agency
EP
USP
sterility
sterile; aseptically filled
form
plate (ready-to-use)
feature
2D bar code
ready-to-use
D
55 mm
pH
7.1 ( in H2O)
application(s)
food and beverages
microbiology
pharmaceutical
surface monitoring
compatibility
for use with Contact Plate
storage temp.
15-25°C
suitability
bacteria
enterobacteriaceae
General description
MacConkey agar is a selective medium for the isolation and cultivation of Enterobacteriaceae, especially E. coli in pharmaceutical, food, and other samples. Enterobacteriaceae can be differentiated even in the presence of Salmonella and Shigella. The pH ranges betweeen 6.9-7.3, which is optimum for the growth of bacteria. The addition of crystal violet and bile salts to MacConkey Agar inhibits the growth of gram-positive bacteria, whereas the growth of most gram-negative bacteria is supported. Bacteria using lactose and producing acids grow in pink to red-colored colonies, and bacteria not degrading lactose will grow colorless. Additionally, E. coli and other acid-forming bacteria will show a zone of precipitated bile salt around the colonies.
Application
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service