Skip to Content
MilliporeSigma

171258

Sigma-Aldrich

AMPK Signaling Activator XV

Synonym(s):

A-769662, 4-Hydroxy-3-(2′-hydroxy-1,1′-biphenyl-4-yl)-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carbonitrile, 4-Hydroxy-3-(2′-hydroxybiphenyl-4-yl)-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carbonitrile, 6,7-Dihydro-4-hydroxy-3-(2′-hydroxy[1,1′-biphenyl]-4-yl)-6-oxothieno[2,3-b]pyridine-5-carbonitrile, A 769662, A769662

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C20H12N2O3S
CAS Number:
Molecular Weight:
360.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

SMILES string

[s]1c2[nH][c](c(c(c2c(c1)c3ccc(cc3)c4c(cccc4)O)O)C#N)=O

InChI

1S/C20H12N2O3S/c21-9-14-18(24)17-15(10-26-20(17)22-19(14)25)12-7-5-11(6-8-12)13-3-1-2-4-16(13)23/h1-8,10,23H,(H2,22,24,25)

Inchi Key

CTESJDQKVOEUOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A-769662 is a potent, β1 subunit-selective, allosteric drug and metabolite (ADaM) site AMPK activator (α1β1γ1 EC50/Emax = 0.15 μM/1.99 vs. 4.51 μM/2.19 with AMP) that promotes a Thr172 phosphorylation in a β1 carbohydrate binding module (CBM) Ser108 phosphorylation-dependent manner. A769662 synergizes with AMP as well as C2 (AMP mimetic) toward Thr172 dephosphorylated/Ser108 phosphorylated AMPK. A-769662 is widely employed in probing AMPK β1 complexes-mediated cellular signaling in cultures (conc range: 1 μM-1 mM) as well as AMPK-dependent physiological and pathological processes in mice and rats in vivo (dosing range: 1-30 mg/kg i.p.).
Potent, β1-selective, allosteric drug and metabolite (ADaM) site AMP-activated protein kinase (AMPK) activator in cultures and in vivo.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service